Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCGTTCGTCCAGGCCAGCCGGG[C/T]AGGCCCCGCCCCCTGGTCCTCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610662 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929134 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101929134 - uncharacterized LOC101929134 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGY - phosphatidylinositol glycan anchor biosynthesis class Y | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042616.2 | 15 | UTR 5 | NP_001036081.1 |
PYURF - PIGY upstream reading frame | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032906.4 | 15 | UTR 5 | NP_116295.1 |