Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGACCCAGAAGTCTTCTGGCGCGG[C/T]GACCAAAAAGGGAGAGCGCGCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC71 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC71 - leucine rich repeat containing 71 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144702.2 | 249 | Missense Mutation | GCG,GTG | A,V 26 | NP_653303.2 | |
XM_005244926.3 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_005244983.1 | |
XM_005244927.3 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_005244984.1 | |
XM_005244928.2 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_005244985.1 | |
XM_006711185.3 | 249 | Intron | XP_006711248.1 | |||
XM_006711186.3 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_006711249.1 | |
XM_006711187.3 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_006711250.1 | |
XM_011509239.2 | 249 | Intron | XP_011507541.1 | |||
XM_011509240.2 | 249 | Intron | XP_011507542.1 | |||
XM_011509241.2 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_011507543.1 | |
XM_017000459.1 | 249 | Missense Mutation | GCG,GTG | A,V 26 | XP_016855948.1 | |
XM_017000460.1 | 249 | Intron | XP_016855949.1 | |||
XM_017000461.1 | 249 | Intron | XP_016855950.1 | |||
XM_017000462.1 | 249 | Intron | XP_016855951.1 |
PEAR1 - platelet endothelial aggregation receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |