Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCTCATACCGCCCACATGCATTA[C/T]GGGATTTGTAGTCCACTCGCGCGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612832 MIM: 607769 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSD17B14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HSD17B14 - hydroxysteroid 17-beta dehydrogenase 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLEKHA4 - pleckstrin homology domain containing A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161354.1 | Intron | NP_001154826.1 | ||||
NM_020904.2 | Intron | NP_065955.2 | ||||
XM_005259107.2 | Intron | XP_005259164.1 | ||||
XM_006723301.2 | Intron | XP_006723364.1 | ||||
XM_011527157.1 | Intron | XP_011525459.1 | ||||
XM_011527158.2 | Intron | XP_011525460.1 | ||||
XM_011527159.1 | Intron | XP_011525461.1 | ||||
XM_011527160.1 | Intron | XP_011525462.1 | ||||
XM_011527162.2 | Intron | XP_011525464.1 | ||||
XM_017027037.1 | Intron | XP_016882526.1 |