Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATCAGAAATTTCAGTAGTGCTG[C/T]GTTGCTATTAGCTTGTATGAGACAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189901 MIM: 611832 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCFC2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GCFC2 - GC-rich sequence DNA-binding factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201334.1 | Intron | NP_001188263.1 | ||||
NM_001201335.1 | Intron | NP_001188264.1 | ||||
NM_003203.4 | Intron | NP_003194.3 | ||||
XM_005264520.3 | Intron | XP_005264577.1 | ||||
XM_011533074.2 | Intron | XP_011531376.1 | ||||
XM_011533075.1 | Intron | XP_011531377.1 | ||||
XM_017004787.1 | Intron | XP_016860276.1 | ||||
XM_017004788.1 | Intron | XP_016860277.1 |
MRPL19 - mitochondrial ribosomal protein L19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |