Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609412 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ERCC8 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ERCC8 - ERCC excision repair 8, CSA ubiquitin ligase complex subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000082.3 | 1215 | UTR 3 | NP_000073.1 | |||
NM_001007233.2 | 1215 | UTR 3 | NP_001007234.1 | |||
NM_001007234.2 | 1215 | Intron | NP_001007235.1 | |||
NM_001290285.1 | 1215 | UTR 3 | NP_001277214.1 |
Set Membership: |
HapMap |