Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGGAAATCCACCAAGTCTCAATA[C/T]AATAATATTCTTTATTCCTGGACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147138 | ||||||||||||||||||||
Literature Links: |
MS4A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MS4A2 - membrane spanning 4-domains A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000139.4 | 61 | UTR 5 | NP_000130.1 | |||
NM_001256916.1 | 61 | UTR 5 | NP_001243845.1 | |||
XM_005273846.4 | 61 | Intron | XP_005273903.1 | |||
XM_011544850.2 | 61 | Intron | XP_011543152.1 | |||
XM_017017362.1 | 61 | Intron | XP_016872851.1 |