Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCCCCGCCCTGCGGCCAGGCA[C/T]ACATGCGGGCACAGGCAGGGGCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601248 | ||||||||||||||||||||
Literature Links: |
BIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BIN1 - bridging integrator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320632.1 | 2306 | UTR 3 | NP_001307561.1 | |||
NM_001320633.1 | 2306 | UTR 3 | NP_001307562.1 | |||
NM_001320634.1 | 2306 | UTR 3 | NP_001307563.1 | |||
NM_001320640.1 | 2306 | UTR 3 | NP_001307569.1 | |||
NM_001320641.1 | 2306 | UTR 3 | NP_001307570.1 | |||
NM_001320642.1 | 2306 | UTR 3 | NP_001307571.1 | |||
NM_004305.3 | 2306 | UTR 3 | NP_004296.1 | |||
NM_139343.2 | 2306 | UTR 3 | NP_647593.1 | |||
NM_139344.2 | 2306 | UTR 3 | NP_647594.1 | |||
NM_139345.2 | 2306 | UTR 3 | NP_647595.1 | |||
NM_139346.2 | 2306 | UTR 3 | NP_647596.1 | |||
NM_139347.2 | 2306 | UTR 3 | NP_647597.1 | |||
NM_139348.2 | 2306 | UTR 3 | NP_647598.1 | |||
NM_139349.2 | 2306 | UTR 3 | NP_647599.1 | |||
NM_139350.2 | 2306 | UTR 3 | NP_647600.1 | |||
NM_139351.2 | 2306 | UTR 3 | NP_647601.1 |