Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602278 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEDD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEDD4 - neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284338.1 | Intron | NP_001271267.1 | ||||
NM_001284339.1 | Intron | NP_001271268.1 | ||||
NM_001284340.1 | Intron | NP_001271269.1 | ||||
NM_006154.3 | Intron | NP_006145.2 | ||||
NM_198400.3 | Intron | NP_940682.2 | ||||
XM_011521624.2 | Intron | XP_011519926.1 | ||||
XM_011521625.2 | Intron | XP_011519927.1 | ||||
XM_011521626.1 | Intron | XP_011519928.1 | ||||
XM_011521627.1 | Intron | XP_011519929.1 |