Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTGCGCCGCTCCTCCAGGTTGTA[A/G]GCTGCTGCCCGCCTTGCCCGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRG2 - neuregulin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184935.1 | 976 | Silent Mutation | GCC,GCT | A,A 496 | NP_001171864.1 | |
NM_004883.2 | 976 | Silent Mutation | GCC,GCT | A,A 562 | NP_004874.1 | |
NM_013981.3 | 976 | Silent Mutation | GCC,GCT | A,A 556 | NP_053584.1 | |
NM_013982.2 | 976 | Silent Mutation | GCC,GCT | A,A 570 | NP_053585.1 | |
NM_013983.2 | 976 | Silent Mutation | GCC,GCT | A,A 564 | NP_053586.1 | |
XM_005268533.3 | 976 | Silent Mutation | GCC,GCT | A,A 171 | XP_005268590.1 | |
XM_006714810.3 | 976 | Intron | XP_006714873.1 | |||
XM_011537713.2 | 976 | Intron | XP_011536015.1 | |||
XM_017010064.1 | 976 | Silent Mutation | GCC,GCT | A,A 182 | XP_016865553.1 | |
XM_017010065.1 | 976 | Silent Mutation | GCC,GCT | A,A 179 | XP_016865554.1 | |
XM_017010066.1 | 976 | Intron | XP_016865555.1 |
PSD2 - pleckstrin and Sec7 domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |