Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGGCACAGGCGGGCGGTCGGC[C/G]GTGTCCTCGCCGTGGGTCACCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124030 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2D6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
CYP2D6*2E,g.997C>G CYP2D6*4A,g.997C>G CYP2D6*4B,g.997C>G CYP2D6*4F,g.997C>G CYP2D6*4G,g.997C>G CYP2D6*4H,g.997C>G CYP2D6*4J,g.997C>G CYP2D6*4L,g.997C>G CYP2D6*4M,g.997C>G CYP2D6*4N,g.997C>G | ||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP2D6 - cytochrome P450 family 2 subfamily D member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000106.5 | 384 | Silent Mutation | ACC,ACG | T,T 98 | NP_000097.3 | |
NM_001025161.2 | 384 | Silent Mutation | ACC,ACG | T,T 98 | NP_001020332.2 | |
XM_011529966.2 | 384 | Intron | XP_011528268.1 | |||
XM_011529968.2 | 384 | Intron | XP_011528270.1 | |||
XM_011529970.2 | 384 | Intron | XP_011528272.1 | |||
XM_011529972.2 | 384 | Intron | XP_011528274.1 |
LOC102723722 - uncharacterized LOC102723722 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017029174.1 | 384 | Intron | XP_016884663.1 |
NDUFA6-AS1 - NDUFA6 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
DME Validated Inventoried |