Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGTGTCCTAGTATGGCATCCC[G/A]TCATTCAGTGCCTGTGTTTGAGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605308 | ||||||||||||||||||||
Literature Links: |
ZNF346 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF346 - zinc finger protein 346 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308213.1 | Intron | NP_001295142.1 | ||||
NM_001308214.1 | Intron | NP_001295143.1 | ||||
NM_001308215.1 | Intron | NP_001295144.1 | ||||
NM_001308216.1 | Intron | NP_001295145.1 | ||||
NM_001308218.1 | Intron | NP_001295147.1 | ||||
NM_001308219.1 | Intron | NP_001295148.1 | ||||
NM_001308221.1 | Intron | NP_001295150.1 | ||||
NM_001308223.1 | Intron | NP_001295152.1 | ||||
NM_012279.3 | Intron | NP_036411.1 |