Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300560 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
PHF8 - PHD finger protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001184896.1 | Intron | NP_001171825.1 | ||||
NM_001184897.1 | Intron | NP_001171826.1 | ||||
NM_001184898.1 | Intron | NP_001171827.1 | ||||
NM_015107.2 | Intron | NP_055922.1 | ||||
XM_005261996.1 | Intron | XP_005262053.1 | ||||
XM_005261997.3 | Intron | XP_005262054.1 | ||||
XM_005261999.1 | Intron | XP_005262056.1 | ||||
XM_005262000.1 | Intron | XP_005262057.1 | ||||
XM_011530778.1 | Intron | XP_011529080.1 | ||||
XM_017029361.1 | Intron | XP_016884850.1 | ||||
XM_017029362.1 | Intron | XP_016884851.1 |