Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCGTTGGGGATGACAGTGCCGTC[A/G]GGCGCCTTCTTGGCTGCAAGACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601670 MIM: 616655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPF1 - double PHD fingers 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135155.2 | 849 | Silent Mutation | CCC,CCT | P,P 291 | NP_001128627.1 | |
NM_001135156.2 | 849 | Silent Mutation | CCC,CCT | P,P 209 | NP_001128628.1 | |
NM_001289978.1 | 849 | Silent Mutation | CCC,CCT | P,P 291 | NP_001276907.1 | |
NM_004647.3 | 849 | Silent Mutation | CCC,CCT | P,P 247 | NP_004638.2 | |
XM_005259289.2 | 849 | Silent Mutation | CCC,CCT | P,P 274 | XP_005259346.1 | |
XM_005259290.3 | 849 | Silent Mutation | CCC,CCT | P,P 265 | XP_005259347.1 | |
XM_005259292.4 | 849 | Silent Mutation | CCC,CCT | P,P 246 | XP_005259349.1 | |
XM_006723407.2 | 849 | Silent Mutation | CCC,CCT | P,P 265 | XP_006723470.1 | |
XM_006723408.3 | 849 | Silent Mutation | CCC,CCT | P,P 263 | XP_006723471.1 | |
XM_006723409.2 | 849 | Silent Mutation | CCC,CCT | P,P 109 | XP_006723472.1 | |
XM_006723410.2 | 849 | Silent Mutation | CCC,CCT | P,P 109 | XP_006723473.1 | |
XM_011527356.1 | 849 | Silent Mutation | CCC,CCT | P,P 274 | XP_011525658.1 | |
XM_011527357.2 | 849 | Silent Mutation | CCC,CCT | P,P 246 | XP_011525659.1 | |
XM_011527358.2 | 849 | Silent Mutation | CCC,CCT | P,P 209 | XP_011525660.1 |
SIPA1L3 - signal induced proliferation associated 1 like 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |