Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCATCACCTCACCTCCTTTAG[A/G]GTAACGATTCCAATGAGTCTGCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600570 | ||||||||||||||||||||
Literature Links: |
CLCN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCN2 - chloride voltage-gated channel 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171087.2 | 2614 | Silent Mutation | ACC,ACT | T,T 814 | NP_001164558.1 | |
NM_001171088.2 | 2614 | Silent Mutation | ACC,ACT | T,T 787 | NP_001164559.1 | |
NM_001171089.2 | 2614 | Intron | NP_001164560.1 | |||
NM_004366.5 | 2614 | Silent Mutation | ACC,ACT | T,T 831 | NP_004357.3 | |
XM_006713489.1 | 2614 | Silent Mutation | ACC,ACT | T,T 796 | XP_006713552.1 | |
XM_006713490.2 | 2614 | Silent Mutation | ACC,ACT | T,T 445 | XP_006713553.1 | |
XM_011512401.1 | 2614 | Intron | XP_011510703.1 |
FAM131A - family with sequence similarity 131 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171093.1 | 2614 | Intron | NP_001164564.1 | |||
NM_144635.4 | 2614 | Intron | NP_653236.3 | |||
XM_005247113.2 | 2614 | Intron | XP_005247170.1 | |||
XM_005247114.3 | 2614 | Intron | XP_005247171.1 | |||
XM_011512413.2 | 2614 | Intron | XP_011510715.1 |