Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTCATTGCAGAAATCACTAAGA[A/T]TTATTCTGTAATCTGCTTACATATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300655 | ||||||||||||||||||||
Literature Links: |
KLHL13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHL13 - kelch like family member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168299.1 | Intron | NP_001161771.1 | ||||
NM_001168300.1 | Intron | NP_001161772.1 | ||||
NM_001168301.1 | Intron | NP_001161773.1 | ||||
NM_001168302.1 | Intron | NP_001161774.1 | ||||
NM_001168303.1 | Intron | NP_001161775.1 | ||||
NM_033495.3 | Intron | NP_277030.2 |