Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAATTACCTGGATGAAACTTTGCAC[A/G]ATTTCAAGAGCAGGTTCAAGAACAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
45 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TRIT1 - tRNA isopentenyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312691.1 | 1040 | Silent Mutation | ATC,ATT | I,I 340 | NP_001299620.1 | |
NM_001312692.1 | 1040 | Silent Mutation | ATC,ATT | I,I 284 | NP_001299621.1 | |
NM_017646.5 | 1040 | Silent Mutation | ATC,ATT | I,I 366 | NP_060116.2 | |
XM_005270954.2 | 1040 | Silent Mutation | ATC,ATT | I,I 285 | XP_005271011.1 | |
XM_006710706.1 | 1040 | Silent Mutation | ATC,ATT | I,I 225 | XP_006710769.1 |
Set Membership: |
HapMap |