Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGCAGGTGCTCCCCGGATGAGA[A/G]TTGAGCTGAGGAGTCCCCTGAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP10B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP10B - ATPase phospholipid transporting 10B (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025153.2 | 4665 | Missense Mutation | CTC,TTC | L,F 1421 | NP_079429.2 | |
XM_006714833.2 | 4665 | Missense Mutation | CTC,TTC | L,F 1421 | XP_006714896.1 | |
XM_011534468.2 | 4665 | Missense Mutation | CTC,TTC | L,F 1421 | XP_011532770.1 | |
XM_011534469.1 | 4665 | Missense Mutation | CTC,TTC | L,F 1421 | XP_011532771.1 | |
XM_011534472.2 | 4665 | Missense Mutation | CTC,TTC | L,F 1393 | XP_011532774.1 | |
XM_017009252.1 | 4665 | Missense Mutation | CTC,TTC | L,F 1409 | XP_016864741.1 | |
XM_017009253.1 | 4665 | Missense Mutation | CTC,TTC | L,F 1354 | XP_016864742.1 |