Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAACATTACTTTCTTTTGTAAAT[A/G]TGGTTTCTACAAAGATGAAACTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 104145 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AFM - afamin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001133.2 | 22 | UTR 5 | NP_001124.1 | |||
XM_017007842.1 | 22 | UTR 5 | XP_016863331.1 | |||
XM_017007843.1 | 22 | UTR 5 | XP_016863332.1 | |||
XM_017007844.1 | 22 | UTR 5 | XP_016863333.1 |