Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604766 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AXDND1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AXDND1 - axonemal dynein light chain domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144696.5 | 971 | Intron | NP_653297.3 | |||
XM_011509165.1 | 971 | Intron | XP_011507467.1 | |||
XM_011509166.2 | 971 | Intron | XP_011507468.1 | |||
XM_011509167.2 | 971 | Intron | XP_011507469.1 | |||
XM_011509168.1 | 971 | Intron | XP_011507470.1 | |||
XM_011509169.1 | 971 | Intron | XP_011507471.1 | |||
XM_011509170.1 | 971 | Intron | XP_011507472.1 | |||
XM_011509171.1 | 971 | Intron | XP_011507473.1 | |||
XM_011509174.1 | 971 | Intron | XP_011507476.1 | |||
XM_011509175.1 | 971 | Intron | XP_011507477.1 | |||
XM_011509176.1 | 971 | Intron | XP_011507478.1 | |||
XM_011509178.2 | 971 | Intron | XP_011507480.1 | |||
XM_011509179.2 | 971 | Intron | XP_011507481.1 | |||
XM_011509180.1 | 971 | Intron | XP_011507482.1 | |||
XM_011509181.2 | 971 | Intron | XP_011507483.1 | |||
XM_017000257.1 | 971 | Intron | XP_016855746.1 | |||
XM_017000258.1 | 971 | Intron | XP_016855747.1 | |||
XM_017000259.1 | 971 | Intron | XP_016855748.1 |
NPHS2 - NPHS2, podocin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297575.1 | 971 | Silent Mutation | CTA,CTG | L,L 278 | NP_001284504.1 | |
NM_014625.3 | 971 | Silent Mutation | CTA,CTG | L,L 346 | NP_055440.1 | |
XM_005245483.3 | 971 | Silent Mutation | CTA,CTG | L,L 287 | XP_005245540.1 | |
XM_017002298.1 | 971 | Silent Mutation | CTA,CTG | L,L 235 | XP_016857787.1 | |
XM_017002299.1 | 971 | Intron | XP_016857788.1 |
Set Membership: |
HapMap JSNP |