Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAAGACAGTATCCAGGAAAACT[G/T]GACTAGAATCTCTTAGCATCTAGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFP69B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFP69B - ZFP69 zinc finger protein B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_023070.2 | Intron | NP_075558.2 | ||||
XM_005271136.1 | Intron | XP_005271193.1 | ||||
XM_005271137.2 | Intron | XP_005271194.1 | ||||
XM_005271138.3 | Intron | XP_005271195.1 | ||||
XM_005271139.3 | Intron | XP_005271196.1 | ||||
XM_017002147.1 | Intron | XP_016857636.1 |