Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 123620 MIM: 123630 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRYBB2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CRYBB2 - crystallin beta B2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CRYBB3 - crystallin beta B3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004076.4 | Intron | NP_004067.1 | ||||
XM_011529902.2 | Intron | XP_011528204.1 | ||||
XM_017028599.1 | Intron | XP_016884088.1 |
KIAA1671 - KIAA1671 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |