Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTCGCTGCAGTAACAGTTCCGC[C/T]GTGACCCACACCCCAAAGGCTCTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602787 MIM: 602791 MIM: 602800 MIM: 602802 MIM: 602818 MIM: 602826 | ||||||||||||||||||||
Literature Links: |
HIST1H2AI PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST1H2AI - histone cluster 1, H2ai | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2AJ - histone cluster 1, H2aj | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021066.2 | Intron | NP_066544.1 |
HIST1H2BL - histone cluster 1, H2bl | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H2BM - histone cluster 1, H2bm | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003521.2 | Intron | NP_003512.1 |
HIST1H3H - histone cluster 1, H3h | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HIST1H4J - histone cluster 1, H4j | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |