Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608473 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANAPC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANAPC1 - anaphase promoting complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022662.3 | Intron | NP_073153.1 | ||||
XM_011511633.2 | Intron | XP_011509935.1 | ||||
XM_011511634.2 | Intron | XP_011509936.1 | ||||
XM_011511636.2 | Intron | XP_011509938.1 | ||||
XM_011511637.2 | Intron | XP_011509939.1 | ||||
XM_017004710.1 | Intron | XP_016860199.1 | ||||
XM_017004711.1 | Intron | XP_016860200.1 | ||||
XM_017004712.1 | Intron | XP_016860201.1 | ||||
XM_017004713.1 | Intron | XP_016860202.1 | ||||
XM_017004714.1 | Intron | XP_016860203.1 | ||||
XM_017004715.1 | Intron | XP_016860204.1 |