Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATGCACCACCTCACTAGGCTA[C/T]AAAGACTGCATGTTATCCATCTACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600583 MIM: 600058 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TEC - tec protein tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003215.2 | Intron | NP_003206.2 | ||||
XM_011513737.2 | Intron | XP_011512039.1 | ||||
XM_011513741.1 | Intron | XP_011512043.1 | ||||
XM_017008569.1 | Intron | XP_016864058.1 |
TXK - TXK tyrosine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |