Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATGCATCCCCAGACAAATGGAA[G/T]GATAAACCAAAAAATCAGTTGAACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616327 | ||||||||||||||||||||
Literature Links: |
CHAMP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHAMP1 - chromosome alignment maintaining phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164144.1 | 608 | Missense Mutation | AAG,AAT | K,N 92 | NP_001157616.1 | |
NM_001164145.1 | 608 | Missense Mutation | AAG,AAT | K,N 92 | NP_001157617.1 | |
NM_032436.2 | 608 | Missense Mutation | AAG,AAT | K,N 92 | NP_115812.1 |
LINC01054 - long intergenic non-protein coding RNA 1054 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |