Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATAAATACCCTCCTTACCTTGCT[C/T]GATTGCCTTGCTGACTACATCATGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYPM PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HYPM - huntingtin interacting protein M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012274.1 | 194 | Silent Mutation | CTC,CTT | L,L 57 | NP_036406.1 |
SYTL5 - synaptotagmin like 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163334.1 | 194 | Intron | NP_001156806.1 | |||
NM_001163335.1 | 194 | Intron | NP_001156807.1 | |||
NM_138780.2 | 194 | Intron | NP_620135.1 | |||
XM_011544001.2 | 194 | Intron | XP_011542303.1 | |||
XM_011544002.2 | 194 | Intron | XP_011542304.1 | |||
XM_017029972.1 | 194 | Intron | XP_016885461.1 | |||
XM_017029973.1 | 194 | Intron | XP_016885462.1 | |||
XM_017029974.1 | 194 | Intron | XP_016885463.1 |
Set Membership: |
HapMap |