Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTBK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTBK1 - tau tubulin kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011514947.2 | 618 | Silent Mutation | TAC,TAT | Y,Y 183 | XP_011513249.1 | |
XM_011514948.2 | 618 | Intron | XP_011513250.1 | |||
XM_017011364.1 | 618 | Silent Mutation | TAC,TAT | Y,Y 183 | XP_016866853.1 | |
XM_017011365.1 | 618 | Silent Mutation | TAC,TAT | Y,Y 183 | XP_016866854.1 | |
XM_017011366.1 | 618 | Silent Mutation | TAC,TAT | Y,Y 183 | XP_016866855.1 | |
XM_017011367.1 | 618 | Silent Mutation | TAC,TAT | Y,Y 183 | XP_016866856.1 | |
XM_017011368.1 | 618 | Intron | XP_016866857.1 |
Set Membership: |
HapMap |