Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF417 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF417 - zinc finger protein 417 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF418 - zinc finger protein 418 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317027.1 | 2172 | Silent Mutation | NP_001303956.1 | |||
NM_001317028.1 | 2172 | Silent Mutation | NP_001303957.1 | |||
NM_001317029.1 | 2172 | Silent Mutation | NP_001303958.1 | |||
NM_001317030.1 | 2172 | Silent Mutation | NP_001303959.1 | |||
NM_133460.2 | 2172 | Silent Mutation | NP_597717.1 | |||
XM_017026304.1 | 2172 | Silent Mutation | XP_016881793.1 | |||
XM_017026305.1 | 2172 | Silent Mutation | XP_016881794.1 | |||
XM_017026306.1 | 2172 | Silent Mutation | XP_016881795.1 | |||
XM_017026307.1 | 2172 | Silent Mutation | XP_016881796.1 | |||
XM_017026308.1 | 2172 | Silent Mutation | XP_016881797.1 | |||
XM_017026309.1 | 2172 | Silent Mutation | XP_016881798.1 | |||
XM_017026310.1 | 2172 | Silent Mutation | XP_016881799.1 | |||
XM_017026311.1 | 2172 | Silent Mutation | XP_016881800.1 | |||
XM_017026312.1 | 2172 | Silent Mutation | XP_016881801.1 | |||
XM_017026313.1 | 2172 | Silent Mutation | XP_016881802.1 | |||
XM_017026314.1 | 2172 | Silent Mutation | XP_016881803.1 |