Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607795 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRPF4 - pre-mRNA processing factor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF183 - ring finger protein 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145051.3 | 1689 | UTR 5 | NP_659488.2 | |||
XM_006716952.2 | 1689 | UTR 5 | XP_006717015.1 | |||
XM_011518218.1 | 1689 | UTR 5 | XP_011516520.1 | |||
XM_011518219.1 | 1689 | UTR 5 | XP_011516521.1 | |||
XM_011518220.1 | 1689 | UTR 5 | XP_011516522.1 | |||
XM_017014268.1 | 1689 | UTR 5 | XP_016869757.1 |