Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGACCCGGTAGCCCGTTCGCTC[C/G]GCGCCGGCGGCCTGTCCCCGCGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D19 - TBC1 domain family member 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001292054.1 | 64 | UTR 5 | NP_001278983.1 | |||
NM_018317.3 | 64 | UTR 5 | NP_060787.2 | |||
XM_011513852.2 | 64 | UTR 5 | XP_011512154.1 | |||
XM_017008363.1 | 64 | UTR 5 | XP_016863852.1 | |||
XM_017008364.1 | 64 | UTR 5 | XP_016863853.1 |