Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGTCACGGTCGCATGTTGTCA[C/T]GAATCCAGTCTAGGTAGTTGGTAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610469 MIM: 173370 | ||||||||||||||||||||
Literature Links: |
AP3M2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AP3M2 - adaptor related protein complex 3 mu 2 subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PLAT - plasminogen activator, tissue type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000930.4 | 1789 | Missense Mutation | CAT,CGT | H,R 557 | NP_000921.1 | |
NM_001319189.1 | 1789 | Missense Mutation | CAT,CGT | H,R 468 | NP_001306118.1 | |
NM_033011.3 | 1789 | Missense Mutation | CAT,CGT | H,R 511 | NP_127509.1 |