Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGAGGTCTGACTTGTTTCCCAC[A/G]AGAACCACAGGTGTATCGTCAGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609591 | ||||||||||||||||||||
Literature Links: |
KIAA0907 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0907 - KIAA0907 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RIT1 - Ras like without CAAX 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256820.1 | 596 | Silent Mutation | CTC,CTT | L,L 95 | NP_001243749.1 | |
NM_001256821.1 | 596 | Silent Mutation | CTC,CTT | L,L 148 | NP_001243750.1 | |
NM_006912.5 | 596 | Silent Mutation | CTC,CTT | L,L 131 | NP_008843.1 |