Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGAGTGAGCAGGAAAAGCAAGAGC[A/G]TCTGGAAACCCAAAGGGAGAAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGES3L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGES3L - prostaglandin E synthase 3 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTGES3L-AARSD1 - PTGES3L-AARSD1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUNDC1 - RUN domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321381.1 | 542 | Missense Mutation | CAT,CGT | H,R 176 | NP_001308310.1 | |
NM_173079.3 | 542 | Missense Mutation | CAT,CGT | H,R 174 | NP_775102.2 | |
XM_005257078.3 | 542 | Missense Mutation | CAT,CGT | H,R 176 | XP_005257135.1 | |
XM_005257080.3 | 542 | Missense Mutation | CAT,CGT | H,R 174 | XP_005257137.1 |