Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACACTTTCCTTTTGCTCGTTCTC[C/T]ATCTAAGAATACAATGTAACAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600073 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LRP2 - LDL receptor related protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004525.2 | 13888 | Missense Mutation | ATA,ATG | I,M 4601 | NP_004516.2 | |
XM_011511183.2 | 13888 | Missense Mutation | ATA,ATG | I,M 4558 | XP_011509485.1 | |
XM_011511184.2 | 13888 | Missense Mutation | ATA,ATG | I,M 3838 | XP_011509486.1 |
Set Membership: |
HapMap |