Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGCCCTTACCTTGAGGAAAAGT[A/G]TAAGCATCTTTAGCATCTTCCTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEMGN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HEMGN - hemogen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018437.4 | 1512 | Silent Mutation | TAC,TAT | Y,Y 449 | NP_060907.2 | |
NM_197978.2 | 1512 | Silent Mutation | TAC,TAT | Y,Y 449 | NP_932095.1 | |
XM_005252086.1 | 1512 | Silent Mutation | TAC,TAT | Y,Y 449 | XP_005252143.1 | |
XM_011518845.2 | 1512 | Silent Mutation | TAC,TAT | Y,Y 449 | XP_011517147.1 | |
XM_011518846.2 | 1512 | Silent Mutation | TAC,TAT | Y,Y 449 | XP_011517148.1 |
TRMO - tRNA methyltransferase O | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |