Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAATGAAGGAGAGAACACTGCTC[G/T]TGGTTTCATAGCCAGCAAAAATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605340 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP3A7 - cytochrome P450 family 3 subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000765.4 | 1036 | Missense Mutation | NP_000756.3 |
CYP3A7-CYP3A51P - CYP3A7-CYP3A51P readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256497.2 | 1036 | Missense Mutation | NP_001243426.2 |
ZSCAN25 - zinc finger and SCAN domain containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145115.2 | 1036 | Intron | NP_660090.2 | |||
XM_005250194.2 | 1036 | Intron | XP_005250251.1 | |||
XM_011515905.2 | 1036 | Intron | XP_011514207.1 | |||
XM_011515906.2 | 1036 | Intron | XP_011514208.1 | |||
XM_011515907.2 | 1036 | Intron | XP_011514209.1 | |||
XM_011515908.2 | 1036 | Intron | XP_011514210.1 | |||
XM_011515909.2 | 1036 | Intron | XP_011514211.1 | |||
XM_011515910.2 | 1036 | Intron | XP_011514212.1 | |||
XM_017011824.1 | 1036 | Intron | XP_016867313.1 | |||
XM_017011825.1 | 1036 | Intron | XP_016867314.1 | |||
XM_017011826.1 | 1036 | Intron | XP_016867315.1 | |||
XM_017011827.1 | 1036 | Intron | XP_016867316.1 |
Set Membership: |
DME Validated Inventoried |