Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACCTTGCCATCACACTCCAAGC[C/T]CCGCCGACGCCGGCGATGAGGGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 147290 | ||||||||||||||||||||
Literature Links: |
INHBA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INHBA - inhibin beta A subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017012174.1 | 2938 | Missense Mutation | XP_016867663.1 | |||
XM_017012175.1 | 2938 | Missense Mutation | XP_016867664.1 | |||
XM_017012176.1 | 2938 | Missense Mutation | XP_016867665.1 |
INHBA-AS1 - INHBA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |