Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGACACTTGAACCCTAGGACCC[A/G]CTTGGGTGGAAGCCTCACCTGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131560 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLOT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FLOT2 - flotillin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004475.2 | Intron | NP_004466.2 | ||||
XM_005257950.3 | Intron | XP_005258007.1 | ||||
XM_005257953.2 | Intron | XP_005258010.1 | ||||
XM_017024394.1 | Intron | XP_016879883.1 | ||||
XM_017024395.1 | Intron | XP_016879884.1 | ||||
XM_017024396.1 | Intron | XP_016879885.1 |