Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTCCCTCAGGGCTGTACCTCAGT[C/T]ACCCCTTCCTGATCCTGCATCCCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 159590 | ||||||||||||||||||||
Literature Links: |
CD33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD33 - CD33 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082618.1 | Intron | NP_001076087.1 | ||||
NM_001177608.1 | Intron | NP_001171079.1 | ||||
NM_001772.3 | Intron | NP_001763.3 | ||||
XM_011527531.2 | Intron | XP_011525833.1 | ||||
XM_011527532.2 | Intron | XP_011525834.1 | ||||
XM_017027508.1 | Intron | XP_016882997.1 | ||||
XM_017027509.1 | Intron | XP_016882998.1 | ||||
XM_017027510.1 | Intron | XP_016882999.1 |
MIR8074 - microRNA 8074 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |