Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGCAGTACTTCTCAGCCTCG[C/G]CCCAGGACATCCCAGAGTGAGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605999 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC10A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLEC10A - C-type lectin domain family 10 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005256412.1 | 946 | Missense Mutation | GCC,GGC | A,G 176 | XP_005256469.1 | |
XM_011523613.1 | 946 | Missense Mutation | GCC,GGC | A,G 206 | XP_011521915.1 | |
XM_011523614.2 | 946 | Missense Mutation | GCC,GGC | A,G 206 | XP_011521916.1 | |
XM_011523615.1 | 946 | Missense Mutation | GCC,GGC | A,G 206 | XP_011521917.1 |