Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATCGGGGGTAAGTAGAGGGATGC[A/G]TCTAATGGCAGCGGCCTCTGGCCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615882 MIM: 611138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR600 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR600 - microRNA 600 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR600HG - MIR600 host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RABGAP1 - RAB GTPase activating protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRBP - spermatid perinuclear RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171137.1 | Intron | NP_001164608.1 | ||||
NM_018387.4 | Intron | NP_060857.2 | ||||
XM_017014890.1 | Intron | XP_016870379.1 | ||||
XM_017014891.1 | Intron | XP_016870380.1 | ||||
XM_017014892.1 | Intron | XP_016870381.1 | ||||
XM_017014893.1 | Intron | XP_016870382.1 | ||||
XM_017014894.1 | Intron | XP_016870383.1 | ||||
XM_017014895.1 | Intron | XP_016870384.1 | ||||
XM_017014896.1 | Intron | XP_016870385.1 | ||||
XM_017014897.1 | Intron | XP_016870386.1 | ||||
XM_017014898.1 | Intron | XP_016870387.1 |