Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTGGGGAGGCTGAGTGCACCG[A/G]CCCCCATGACCCACAGCCGTTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613593 MIM: 613594 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BTN3A1 - butyrophilin subfamily 3 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145008.1 | Intron | NP_001138480.1 | ||||
NM_001145009.1 | Intron | NP_001138481.1 | ||||
NM_007048.5 | Intron | NP_008979.3 | ||||
NM_194441.2 | Intron | NP_919423.1 | ||||
XM_005248833.2 | Intron | XP_005248890.1 | ||||
XM_005248834.3 | Intron | XP_005248891.1 | ||||
XM_006714983.2 | Intron | XP_006715046.1 |