Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATCAGGTAATAAATTAACCAG[A/G]CAGGTTCACATTCAATCAGATAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103740 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADH4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADH4 - alcohol dehydrogenase 4 (class II), pi polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000670.4 | 1407 | UTR 3 | NP_000661.2 | |||
NM_001306171.1 | 1407 | UTR 3 | NP_001293100.1 | |||
NM_001306172.1 | 1407 | UTR 3 | NP_001293101.1 | |||
XM_017007713.1 | 1407 | Intron | XP_016863202.1 | |||
XM_017007714.1 | 1407 | Intron | XP_016863203.1 | |||
XM_017007715.1 | 1407 | Intron | XP_016863204.1 |
LOC100507053 - uncharacterized LOC100507053 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |