Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACCAGAGCCCACCGCACAGGCA[A/G]TTCTTGATCCAGCGCTGCTCCCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134350 MIM: 604062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFD - complement factor D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MED16 - mediator complex subunit 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005481.2 | 2629 | Silent Mutation | AAC,AAT | N,N 826 | NP_005472.2 | |
XM_017026120.1 | 2629 | Silent Mutation | AAC,AAT | N,N 757 | XP_016881609.1 | |
XM_017026121.1 | 2629 | Silent Mutation | AAC,AAT | N,N 651 | XP_016881610.1 |