Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTCAAATTCACCCTTGAACATA[A/C]GTGGCAAATTCTTGAGACTTGTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612471 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AGXT2 - alanine--glyoxylate aminotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306173.1 | 1884 | Intron | NP_001293102.1 | |||
NM_031900.3 | 1884 | UTR 3 | NP_114106.1 | |||
XM_005248337.2 | 1884 | UTR 3 | XP_005248394.1 | |||
XM_005248338.2 | 1884 | UTR 3 | XP_005248395.1 | |||
XM_017009748.1 | 1884 | UTR 3 | XP_016865237.1 |