Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCCATCTTTAAATTAATGATTCC[C/G]AATATTACCCAGTACAGTCCTCACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDHAL6A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LDHAL6A - lactate dehydrogenase A like 6A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144071.1 | 455 | Silent Mutation | CCC,CCG | P,P 122 | NP_001137543.1 | |
NM_144972.4 | 455 | Silent Mutation | CCC,CCG | P,P 122 | NP_659409.2 | |
XM_005252805.4 | 455 | Silent Mutation | CCC,CCG | P,P 146 | XP_005252862.2 | |
XM_011519922.2 | 455 | Silent Mutation | CCC,CCG | P,P 146 | XP_011518224.1 | |
XM_011519923.2 | 455 | Silent Mutation | CCC,CCG | P,P 122 | XP_011518225.1 | |
XM_011519924.2 | 455 | Silent Mutation | CCC,CCG | P,P 122 | XP_011518226.1 | |
XM_011519925.1 | 455 | Silent Mutation | CCC,CCG | P,P 122 | XP_011518227.1 |