Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCCCTGCGGGTGACAGGCTGTC[C/T]GGGCCCCACATACCTCTGTCTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616062 MIM: 614939 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKLE2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ANKLE2 - ankyrin repeat and LEM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015114.2 | Intron | NP_055929.1 | ||||
XM_005266159.3 | Intron | XP_005266216.1 | ||||
XM_005266160.1 | Intron | XP_005266217.1 | ||||
XM_006719735.1 | Intron | XP_006719798.1 | ||||
XM_011534787.2 | Intron | XP_011533089.1 | ||||
XM_011534788.1 | Intron | XP_011533090.1 | ||||
XM_017019076.1 | Intron | XP_016874565.1 |
PGAM5 - PGAM family member 5, mitochondrial serine/threonine protein phosphatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |