Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPS6KC1 - ribosomal protein S6 kinase C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136138.2 | 126 | UTR 5 | NP_001129610.1 | |||
NM_001287218.1 | 126 | UTR 5 | NP_001274147.1 | |||
NM_001287219.1 | 126 | UTR 5 | NP_001274148.1 | |||
NM_001287220.1 | 126 | UTR 5 | NP_001274149.1 | |||
NM_001287221.1 | 126 | UTR 5 | NP_001274150.1 | |||
NM_012424.4 | 126 | UTR 5 | NP_036556.2 | |||
XM_005273095.1 | 126 | UTR 5 | XP_005273152.1 | |||
XM_005273096.1 | 126 | UTR 5 | XP_005273153.1 | |||
XM_006711272.1 | 126 | UTR 5 | XP_006711335.1 | |||
XM_017001024.1 | 126 | Intron | XP_016856513.1 | |||
XM_017001025.1 | 126 | UTR 5 | XP_016856514.1 | |||
XM_017001026.1 | 126 | UTR 5 | XP_016856515.1 | |||
XM_017001027.1 | 126 | UTR 5 | XP_016856516.1 | |||
XM_017001028.1 | 126 | UTR 5 | XP_016856517.1 | |||
XM_017001029.1 | 126 | UTR 5 | XP_016856518.1 | |||
XM_017001030.1 | 126 | UTR 5 | XP_016856519.1 | |||
XM_017001031.1 | 126 | UTR 5 | XP_016856520.1 | |||
XM_017001032.1 | 126 | UTR 5 | XP_016856521.1 | |||
XM_017001033.1 | 126 | UTR 5 | XP_016856522.1 | |||
XM_017001034.1 | 126 | UTR 5 | XP_016856523.1 |
Set Membership: |
HapMap |