Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCTCTTCTGCCCTATTTTGGACC[A/G]GGAGATGAGCAATGTTCTTACTTCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616880 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D22B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TBC1D22B - TBC1 domain family member 22B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017772.3 | Intron | NP_060242.2 | ||||
XM_011514738.2 | Intron | XP_011513040.1 | ||||
XM_011514739.1 | Intron | XP_011513041.1 |
Set Membership: |
HapMap |