Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTTACTGACAGGTTGCCCACCT[C/T]CCCCAACGCCACCCCGCTTCGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616906 MIM: 190070 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASC1 - cancer susceptibility candidate 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082972.2 | Intron | NP_001076441.1 | ||||
NM_001082973.2 | Intron | NP_001076442.1 | ||||
NM_001204101.2 | Intron | NP_001191030.1 | ||||
NM_001204102.2 | Intron | NP_001191031.1 | ||||
NM_001319977.1 | Intron | NP_001306906.1 | ||||
NM_001319978.1 | Intron | NP_001306907.1 | ||||
NM_018272.4 | Intron | NP_060742.3 | ||||
XM_005253406.1 | Intron | XP_005253463.1 | ||||
XM_005253408.1 | Intron | XP_005253465.1 | ||||
XM_005253409.1 | Intron | XP_005253466.1 | ||||
XM_006719101.1 | Intron | XP_006719164.1 | ||||
XM_011520723.1 | Intron | XP_011519025.1 | ||||
XM_011520726.1 | Intron | XP_011519028.1 | ||||
XM_011520727.1 | Intron | XP_011519029.1 | ||||
XM_017019560.1 | Intron | XP_016875049.1 | ||||
XM_017019561.1 | Intron | XP_016875050.1 | ||||
XM_017019562.1 | Intron | XP_016875051.1 | ||||
XM_017019563.1 | Intron | XP_016875052.1 | ||||
XM_017019564.1 | Intron | XP_016875053.1 | ||||
XM_017019565.1 | Intron | XP_016875054.1 | ||||
XM_017019566.1 | Intron | XP_016875055.1 | ||||
XM_017019567.1 | Intron | XP_016875056.1 | ||||
XM_017019568.1 | Intron | XP_016875057.1 |
KRAS - KRAS proto-oncogene, GTPase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LYRM5 - LYR motif containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001660.2 | Intron | NP_001001660.2 | ||||
XM_005253319.4 | Intron | XP_005253376.1 | ||||
XM_005253320.4 | Intron | XP_005253377.1 | ||||
XM_017018850.1 | Intron | XP_016874339.1 |